Anti-LY9 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-2529R-A488
10341-088EA 1226.35 CAD
10341-088
Anti-LY9 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
May participate in adhesion reactions between T lymphocytes and accessory cells by homophilic interaction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LY9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR