Anti-LY9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-2529R-HRP
10341-106EA 1057.62 CAD
10341-106
Anti-LY9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May participate in adhesion reactions between T lymphocytes and accessory cells by homophilic interaction.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: LY9
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR