Anti-KLK10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2531R-A555
10340-890EA 1226.35 CAD
10340-890
Anti-KLK10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Has a tumor-suppressor role for NES1 in breast and prostate cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLK10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR