Anti-KLK10 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2531R-CY3
10340-896EA 1114.03 CAD
10340-896
Anti-KLK10 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Has a tumor-suppressor role for NES1 in breast and prostate cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLK10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR