Anti-VCAN Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2533R-CY7
10341-320EA 1226.35 CAD
10341-320
Anti-VCAN Rabbit Polyclonal Antibody (Cy7®)
Antibodies
May play a role in intercellular signaling and in connecting cells with the extracellular matrix. May take part in the regulation of cell motility, growth and differentiation. Binds hyaluronic acid.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VCAN
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR