Anti-VCAN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-2533R-HRP
10341-324EA 1226.35 CAD
10341-324
Anti-VCAN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May play a role in intercellular signaling and in connecting cells with the extracellular matrix. May take part in the regulation of cell motility, growth and differentiation. Binds hyaluronic acid.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: VCAN
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR