Anti-CTF1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2534R-CY5.5
10341-144EA 1057.62 CAD
10341-144
Anti-CTF1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Induces cardiac myocyte hypertrophy in vitro. Binds to and activates the ILST/gp13 receptor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR