Anti-CTF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-2534R-FITC
10341-148EA 1114.03 CAD
10341-148
Anti-CTF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Induces cardiac myocyte hypertrophy in vitro. Binds to and activates the ILST/gp13 receptor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTF1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR