Anti-CTF1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-2534R-HRP
10341-150EA 1226.35 CAD
10341-150
Anti-CTF1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Induces cardiac myocyte hypertrophy in vitro. Binds to and activates the ILST/gp13 receptor.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CTF1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR