Anti-COLEC12 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-2541R-CY5
10341-282EA 961.37 CAD
10341-282
Anti-COLEC12 Rabbit Polyclonal Antibody (Cy5®)
Scavenger receptor that displays several functions associated with host defense. Promotes binding and phagocytosis of Gram-positive, Gram-negative bacteria and yeast. Mediates the recognition, internalization and degradation of oxidatively modified low density lipoprotein (oxLDL) by vascular endothelial cells. Binds to several carbohydrates including Gal-type ligands, D-galactose, L- and D-fucose, GalNAc, T and Tn antigens in a calcium-dependent manner and internalizes specifically GalNAc in nurse-like cells. Binds also to sialyl Lewis X or a trisaccharide and asialo-orosomucoid (ASOR). May also play a role in the clearance of amyloid beta in Alzheimer disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COLEC12
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR