Anti-CLEC1B Rabbit Polyclonal Antibody

Supplier: Bioss
BS-2543R
10341-332EA 830.46 CAD
10341-332
Anti-CLEC1B Rabbit Polyclonal Antibody
Antibodies
Natural killer (NK) cells express multiple C-type (calcium-dependent) lectin-like receptors, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC1B (also known as CLEC-2) is a C-type lectin-like receptor expressed in myeloid cells and NK cells.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLEC1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR