Anti-CXCL5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-2549R-FITC
10341-508EA 1057.62 CAD
10341-508
Anti-CXCL5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
May participate in the recruitment of inflammatory cells by injured or infected tissue. GCP-2(1-78) and, more potent, GCP-2(9-78) attract neutrophils and are involved in neutrophil activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXCL5
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR