Anti-CXCL9 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2551R-CY3
10341-350EA 1114.03 CAD
10341-350
Anti-CXCL9 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Cytokine that affects the growth, movement, or activation state of cells that participate in immune and inflammatory response. Chemotactic for activated T-cells. Binds to CXCR3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXCL9
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR