Anti-CXCL11 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2552R-A350
10341-472EA 1114.03 CAD
10341-472
Anti-CXCL11 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Chemotactic for interleukin-activated T-cells but not unstimulated T-cells, neutrophils or monocytes. Induces calcium release in activated T-cells. Binds to CXCR3. May play an important role in CNS diseases which involve T-cell recruitment. May play a role in skin immune responses (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXCL11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR