Anti-CXCL13 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2553R-CY7
10341-572EA 1226.35 CAD
10341-572
Anti-CXCL13 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Chemotactic for B-lymphocytes but not for T-lymphocytes, monocytes and neutrophils. Does not induce calcium release in B-lymphocytes. Binds to BLR1/CXCR5.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXCL13
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR