Anti-CXCL15 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2554R-A555
10341-584EA 1226.35 CAD
10341-584
Anti-CXCL15 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
CXCL15 (Lungkine) is a CXC chemokine that is expressed in murine lung epithelial cells and, to a lesser extent, in certain fetal tissues. No human homolog has been identified and a specific cell surface receptor has not yet been found. Lungkine expression in lung tissue is elevated in response to inflammation, at which time it acts to specifically recruit neutrophils and direct them into the lung airway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXCL15
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR