Anti-LAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-2556R-A647
10341-634EA 1057.62 CAD
10341-634
Anti-LAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
May play a role in dendritic cell function and in adaptive immunity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LAMP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR