Anti-LAMP3 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-2556R-CY5
10341-640EA 1226.35 CAD
10341-640
Anti-LAMP3 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
May play a role in dendritic cell function and in adaptive immunity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LAMP3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR