Anti-LY75 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-2565R-A555
10341-664EA 1179.34 CAD
10341-664
Anti-LY75 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Acts as an endocytic receptor to direct captured antigens from the extracellular space to a specialized antigen-processing compartment (By similarity). Causes reduced proliferation of B-lymphocytes.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LY75
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR