Anti-CLEC6A Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-2566R-A647
10341-840EA 1114.03 CAD
10341-840
Anti-CLEC6A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Dectin 2 is a type II transmembrane protein that is a member of the C type lectin superfamily. In mouse, dectin 2 is predominantly expressed on tissue macrophages and some dendritic cells. Significant expression of Dectin 2 has been reported on macrophages in the red pulp and marginal zones of the spleen, kupffer cells in the liver and alveolar macrophages in the lung. Peripheral blood monocytes express low levels of dectin 2 but transient up regulation of expression has been demonstrated on monocytes at sites of inflammation. The function of dectin 2 has not been fully determined but studies suggest a possible role in immune surveillance.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLEC6A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR