Anti-FCGR2A Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-2573R-A647
10341-700EA 1226.35 CAD
10341-700
Anti-FCGR2A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Binds to the Fc region of immunoglobulins gamma. Low affinity receptor. By binding to IgG it initiates cellular responses against pathogens and soluble antigens. Promotes phagocytosis of opsonized antigens.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD32
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR