Anti-CSF3R Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-2574R-A488
10341-878EA 1179.34 CAD
10341-878
Anti-CSF3R Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Receptor for granulocyte colony-stimulating factor (CSF3), essential for granulocytic maturation. Plays a crucial role in the proliferation, differientation and survival of cells along the neutrophilic lineage. In addition it may function in some adhesion or recognition events at the cell surface.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSF3R
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR