Anti-CSF3R Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2574R-CY5.5
10341-890EA 1179.34 CAD
10341-890
Anti-CSF3R Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Receptor for granulocyte colony-stimulating factor (CSF3), essential for granulocytic maturation. Plays a crucial role in the proliferation, differientation and survival of cells along the neutrophilic lineage. In addition it may function in some adhesion or recognition events at the cell surface.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSF3R
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR