Anti-GZMA Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-2578R-A647
10342-054EA 1057.62 CAD
10342-054
Anti-GZMA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Abundant protease in the cytosolic granules of cytotoxic T-cells and NK-cells which activates caspase-independent cell death with morphological features of apoptosis when delivered into the target cell through the immunological synapse. It cleaves after Lys or Arg. Cleaves APEX1 after 'Lys-31' and destroys its oxidative repair activity. Cleaves the nucleosome assembly protein SET after 'Lys-189', which disrupts its nucleosome assembly activity and allows the SET complex to translocate into the nucleus to nick and degrade the DNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GZMA/Granzyme A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR