Anti-GZMH Rabbit Polyclonal Antibody

Supplier: Bioss
BS-2579R
10342-118EA 906.49 CAD
10342-118
Anti-GZMH Rabbit Polyclonal Antibody
Antibodies
protease required for lymphocyte cytotoxic granules to lyse target cells [RGD]. Granzyme B is a cysteine protease found in the cytoplasmic lysosome-like granules of cytolytic T lymphocytes (CTL) and natural killer (NK) cells and is is required for the induction of target cell lysis. This occurs as part of cell mediated immune responses, and can activate apoptosis in target cells by both caspase dependent and caspase independent mechanisms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Granzyme D/B/E/N/G/F/H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR