Anti-GZMH Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-2579R-A555
10342-124EA 1179.34 CAD
10342-124
Anti-GZMH Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
protease required for lymphocyte cytotoxic granules to lyse target cells [RGD]. Granzyme B is a cysteine protease found in the cytoplasmic lysosome-like granules of cytolytic T lymphocytes (CTL) and natural killer (NK) cells and is is required for the induction of target cell lysis. This occurs as part of cell mediated immune responses, and can activate apoptosis in target cells by both caspase dependent and caspase independent mechanisms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Granzyme D/B/E/N/G/F/H
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR