Anti-IGSF8 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2584R-CY5.5
10342-164EA 1057.29 CAD
10342-164
Anti-IGSF8 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
May play a key role in diverse functions ascribed to CD81 and CD9 such as oocytes fertilization or hepatitis C virus function. May regulate proliferation and differentiation of keratinocytes. May be a negative regulator of cell motility: suppresses T-cell mobility coordinately with CD81, associates with CD82 to suppress prostate cancer cell migration, regulates epidermoid cell reaggregation and motility on laminin-5 with CD9 and CD81 as key linkers. May also play a role on integrin-dependent morphology and motility functions. May participate in the regulation of neurite outgrowth and maintenance of the neural network in the adult brain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGSF8
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR