Anti-FBXO32 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-2591R-CY7
10341-864EA 1114.03 CAD
10341-864
Anti-FBXO32 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. Probably recognizes and binds to phosphorylated target proteins during skeletal muscle atrophy. Recognizes TERF1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Fbx32
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR