Anti-IL3 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-2598R-CY5.5
10342-472EA 1226.35 CAD
10342-472
Anti-IL3 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Granulocyte/macrophage colony-stimulating factors are cytokines that act in hematopoiesis by controlling the production, differentiation, and function of 2 related white cell populations of the blood, the granulocytes and the monocytes-macrophages. This CSF induces granulocytes, macrophages, mast cells, stem cells, erythroid cells, eosinophils and megakaryocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR