Anti-IL5RA Rabbit Polyclonal Antibody (PE (Phycoerythrin))

BS-2601R-PE
10342-412EA 606.5 CAD
10342-412
Anti-IL5RA Rabbit Polyclonal Antibody (PE (Phycoerythrin))
The protein encoded by this gene is an interleukin 5 specific subunit of a heterodimeric cytokine receptor. The receptor is comprised of a ligand specific alpha subunit and a signal transducing beta subunit shared by the receptors for interleukin 3(IL3), colony stimulating factor 2 (CSF2/GM-CSF), and interleukin 5(IL5). The binding of this protein to IL5 depends on the beta subunit. The beta subunit is activated by the ligand binding, and is required for the biological activities of IL5. This protein has been found to interact with syndecan binding protein (syntenin), which is required for IL5 mediated activation of the transcription factor SOX4. Six alternatively spliced transcript variants encoding three distinct isoforms have been reported.

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: IL5RA
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR