Anti-IL12RB2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-2604R-A350
10342-568EA 1225.99 CAD
10342-568
Anti-IL12RB2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Receptor for interleukin-12. This subunit is the signaling component coupling to the JAK2/STAT4 pathway. Promotes the proliferation of T-cells as well as NK cells. Induces the promotion of T-cells towards the Th1 phenotype by strongly enhancing IFN-gamma production.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL12RB2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR