Anti-IL17RA Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2606R-A350
10342-618EA 1114.03 CAD
10342-618
Anti-IL17RA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Receptor for IL17A, IL17F and, in dimer with IL17RE, for IL17C. Binds its IL17A ligand with low affinity, suggesting that additional components are involved in IL17A-induced signaling.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR