Anti-IL17RA Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2606R-CY3
10342-628EA 1226.35 CAD
10342-628
Anti-IL17RA Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Receptor for IL17A, IL17F and, in dimer with IL17RE, for IL17C. Binds its IL17A ligand with low affinity, suggesting that additional components are involved in IL17A-induced signaling.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR