Anti-IL17RA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-2606R-HRP
10342-638EA 1114.03 CAD
10342-638
Anti-IL17RA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Receptor for IL17A, IL17F and, in dimer with IL17RE, for IL17C. Binds its IL17A ligand with low affinity, suggesting that additional components are involved in IL17A-induced signaling.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR