Anti-IL17RC Rabbit Polyclonal Antibody

Supplier: Bioss
BS-2607R
10342-642EA 874.75 CAD
10342-642
Anti-IL17RC Rabbit Polyclonal Antibody
Antibodies
IL17RC is a single pass transmembrane protein that shares limited similarity with the interleukin 17 receptor. It is widely expressed in non leukocyte tissues and forms a heteromeric receptor for IL17 with IL17RA. At least four different isoforms are known to exist.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL17RC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR