Anti-IL17B Rabbit Polyclonal Antibody (Biotin)

BS-2609R-BIO
10342-722EA 793.94 CAD
10342-722
Anti-IL17B Rabbit Polyclonal Antibody (Biotin)
Stimulates the release of tumor necrosis factor alpha and IL-1-beta from the monocytic cell line THP-1.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: IL17B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR