Anti-IL17B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-2609R-FITC
10342-732EA 1114.03 CAD
10342-732
Anti-IL17B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Stimulates the release of tumor necrosis factor alpha and IL-1-beta from the monocytic cell line THP-1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL17B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR