Anti-IL17RB Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2610R-A350
10342-738EA 1114.03 CAD
10342-738
Anti-IL17RB Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Receptor for the proinflammatory cytokines IL17B and IL17E. May play a role in controlling the growth and/or differentiation of hematopoietic cells.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL17RB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR