Anti-IL18RAP Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-2616R-CY7
10342-854EA 1057.62 CAD
10342-854
Anti-IL18RAP Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Required for the high affinity binding of interleukin 18 (IL-18) to its receptor complex (By similarity). Together with IL18R1 mediates IL-18-dependent activation of NF-kappa-B and JNK.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL18RAP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR