Anti-IL20RA Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-2619R-CY5
10342-956EA 1012.64 CAD
10342-956
Anti-IL20RA Rabbit Polyclonal Antibody (Cy5®)
The IL2RA/IL2RB dimer is a receptor for IL19, IL2 and IL24. The IL2RA/IL1RB dimer is a receptor for IL26.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL20RA
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR