Anti-IL20RB Rabbit Polyclonal Antibody

Supplier: Bioss

BS-2620R
10342-966EA 982.3 CAD
10342-966
Anti-IL20RB Rabbit Polyclonal Antibody
Antibodies
The IL20RA/IL20RB dimer is a receptor for IL19, IL20 and IL24. IL20RB belongs to the type II cytokine receptor family. It is widely expressed, with highest levels in skin and testis; highly expressed in psoriatic skin.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL20RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR