Anti-IL21R Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-2622R-CY7
10343-066EA 1057.62 CAD
10343-066
Anti-IL21R Rabbit Polyclonal Antibody (Cy7®)
Antibodies
This is a receptor for interleukin-21.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL21R
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR