Anti-IL21R Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-2622R-FITC
10343-068EA 1057.62 CAD
10343-068
Anti-IL21R Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
This is a receptor for interleukin-21.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL21R
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR