Anti-IL22 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2623R-CY3
10343-084EA 1114.03 CAD
10343-084
Anti-IL22 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Cytokine that contributes to the inflammatory response in vivo.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL22
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR