Anti-PROK1/PRK1/EG-VEGF Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2638R-CY5.5
10343-322EA 1114.03 CAD
10343-322
Anti-PROK1/PRK1/EG-VEGF Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Potently contracts gastrointestinal (GI) smooth muscle. Induces proliferation, migration and fenestration (the formation of membrane discontinuities) in capillary endothelial cells derived from endocrine glands. Has little or no effect on a variety of other endothelial and non-endothelial cell types. Induces proliferation and differentiation, but not migration, of enteric neural crest cells. Directly influences neuroblastoma progression by promoting the proliferation and migration of neuroblastoma cells. Positively regulates PTGS2 expression and prostaglandin synthesis. May play a role in placentation. May play a role in normal and pathological testis angiogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PROK1/PRK1/EG-VEGF
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR