Anti-PTGES2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2639R-CY3
10343-440EA 1226.35 CAD
10343-440
Anti-PTGES2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Isomerase that catalyzes the conversion of PGH2 into the more stable prostaglandin E2 (PGE2) (By similarity). May also have transactivation activity toward IFN-gamma (IFNG), possibly via an interaction with CEBPB; however, the relevance of transcription activation activity remains unclear.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTGES2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR