Anti-PTGES2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-2639R-HRP
10343-450EA 1226.35 CAD
10343-450
Anti-PTGES2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Isomerase that catalyzes the conversion of PGH2 into the more stable prostaglandin E2 (PGE2) (By similarity). May also have transactivation activity toward IFN-gamma (IFNG), possibly via an interaction with CEBPB; however, the relevance of transcription activation activity remains unclear.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PTGES2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR