Anti-ITGA6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-2641R-A555
10343-352EA 1114.03 CAD
10343-352
Anti-ITGA6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Integrin alpha-6/beta-1 is a receptor for laminin on platelets. Integrin alpha-6/beta-4 is a receptor for laminin in epithelial cells and it plays a critical structural role in the hemidesmosome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITGA6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR