Anti-KIR2DL3 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-2642R-CY5
10343-384EA 1226.35 CAD
10343-384
Anti-KIR2DL3 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Receptor on natural killer (NK) cells for HLA-C alleles (HLA-Cw1, HLA-Cw3 and HLA-Cw7). Inhibits the activity of NK cells thus preventing cell lysis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIR2DL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR