Anti-KIR2DL4 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2643R-CY5.5
10343-038EA 1057.62 CAD
10343-038
Anti-KIR2DL4 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Receptor on natural killer (NK) cells for HLA-C alleles. Inhibits the activity of NK cells thus preventing cell lysis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIR2DL4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR